Merck
  • A high-coverage shRNA screen identifies TMEM129 as an E3 ligase involved in ER-associated protein degradation.
A high-coverage shRNA screen identifies TMEM129 as an E3 ligase involved in ER-associated protein degradation.
Nature communications (2014-05-09)
Michael L van de Weijer, Michael C Bassik, Rutger D Luteijn, Cornelia M Voorburg, Mirjam A M Lohuis, Elisabeth Kremmer, Rob C Hoeben, Emily M LeProust, Siyuan Chen, Hanneke Hoelen, Maaike E Ressing, Weronika Patena, Jonathan S Weissman, Michael T McManus, Emmanuel J H J Wiertz, Robert Jan Lebbink
ABSTRACT
Misfolded ER proteins are retrotranslocated into the cytosol for degradation via the ubiquitin-proteasome system. The human cytomegalovirus protein US11 exploits this ER-associated protein degradation (ERAD) pathway to downregulate HLA class I molecules in virus-infected cells, thereby evading elimination by cytotoxic T-lymphocytes. US11-mediated degradation of HLA class I has been instrumental in the identification of key components of mammalian ERAD, including Derlin-1, p97, VIMP and SEL1L. Despite this, the process governing retrotranslocation of the substrate is still poorly understood. Here using a high-coverage genome-wide shRNA library, we identify the uncharacterized protein TMEM129 and the ubiquitin-conjugating E2 enzyme UBE2J2 to be essential for US11-mediated HLA class I downregulation. TMEM129 is an unconventional C4C4-type RING finger E3 ubiquitin ligase that resides within a complex containing various other ERAD components, including Derlin-1, Derlin-2, VIMP and p97, indicating that TMEM129 is an integral part of the ER-resident dislocation complex mediating US11-induced HLA class I degradation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse