Merck
Guanidine methods for total RNA preparation.
Current protocols in molecular biology (2008-02-12)
R E Kingston, P Chomczynski, N Sacchi
ABSTRACT
Three different methods for RNA preparation using guanidine are presented in this unit--a single-step isolation method employing liquid-phase separation to selectively extract total RNA from tissues and cultured cells and two methods that rely on a CsCl step gradient to isolate total RNA.
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%