Merck
  • Enantiomeric impurities in chiral synthons, catalysts, and auxiliaries. Part 3.
Enantiomeric impurities in chiral synthons, catalysts, and auxiliaries. Part 3.
Tetrahedron, asymmetry (2006-10-27)
Ke Huang, Zachary S Breitbach, Daniel W Armstrong
ABSTRACT
The enantiomeric excess of chiral reagents used in asymmetric syntheses directly affects the reaction selectivity and product purity. In this work, 84 of the more recently available chiral compounds were evaluated to determine their actual enantiomeric composition. These compounds are widely used in asymmetric syntheses as chiral synthons, catalysts, and auxiliaries. These include chiral alcohols, amines, amino alcohols, amides, carboxylic acids, epoxides, esters, ketones, and oxolanes among other classes of compounds. All enantiomeric test results were categorized within five purity levels (i.e. <0.01%, 0.01% to 0.1%, 0.1% to 1%, 1% to 10%, and >10%). The majority of the reagents tested were determined to have enantiomeric impurities over 0.01%, and two of them were found to contain enantiomeric impurities exceeding the 10% level. The most effective enantioselective analysis method was a GC approach using a Chiraldex GTA chiral stationary phase (CSP). This method worked exceedingly well with chiral amines and alcohols.
Product Description
Supelco
Astec® CHIRALDEX GC Column Screening Kit, L × I.D. 30 m × 0.25 mm, pkg of 1 kit
Supelco
Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 20 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 50 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX B-DM Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX B-TA Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX B-DM Capillary GC Column, L × I.D. 50 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-BP Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-PN Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm