Merck
  • Mesalamine did not prevent recurrent diverticulitis in phase 3 controlled trials.
Mesalamine did not prevent recurrent diverticulitis in phase 3 controlled trials.
Gastroenterology (2014-07-20)
Jeffrey B Raskin, Michael A Kamm, M Mazen Jamal, Juan Márquez, Ehud Melzer, Robert E Schoen, Tibor Szalóki, Karen Barrett, Paul Streck
ABSTRACT
No therapy has been proven to prevent the recurrence of diverticulitis. Mesalamine has shown efficacy in preventing relapse in inflammatory bowel disease, and there is preliminary evidence that it might be effective for diverticular disease. We investigated the efficacy of mesalamine in preventing recurrence of diverticulitis in 2 identical but separate phase 3, randomized, double-blind, placebo-controlled, multicenter trials (identical confirmatory trials were conducted for regulatory reasons). We evaluated the efficacy and safety of multimatrix mesalamine vs placebo in the prevention of recurrent diverticulitis in 590 (PREVENT1) and 592 (PREVENT2) adult patients with ≥1 episodes of acute diverticulitis in the previous 24 months that resolved without surgery. Patients received mesalamine (1.2 g, 2.4 g, or 4.8 g) or placebo once daily for 104 weeks. The primary end point was the proportion of recurrence-free patients at week 104. Diverticulitis recurrence was defined as surgical intervention at any time for diverticular disease or presence of computed tomography scan results demonstrating bowel wall thickening (>5 mm) and/or fat stranding consistent with diverticulitis. For a portion of the study, recurrence also required the presence of abdominal pain and an increase in white blood cells. Mesalamine did not reduce the rate of diverticulitis recurrence at week 104. Among patients in PREVENT1, 53%-63% did not have disease recurrence, compared with 65% of those given placebo. Among patients in PREVENT2, 59%-69% of patients did not have disease recurrence, compared with 68% of those given placebo. Mesalamine did not reduce time to recurrence, and the proportions of patients requiring surgery were comparable among treatment groups. No new adverse events were identified with mesalamine administration. Mesalamine was not superior to placebo in preventing recurrent diverticulitis. Mesalamine is not recommended for this indication. ClinicalTrials.gov ID: NCT00545740 and NCT00545103.
Product Description
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%