Merck
  • Intravenous lidocaine decreases tumor necrosis factor alpha expression both locally and systemically in pigs undergoing lung resection surgery.
Intravenous lidocaine decreases tumor necrosis factor alpha expression both locally and systemically in pigs undergoing lung resection surgery.
Anesthesia and analgesia (2014-07-19)
Ignacio Garutti, Lisa Rancan, Carlos Simón, Gabriel Cusati, Guillermo Sanchez-Pedrosa, Francisco Moraga, Luis Olmedilla, Maria Teresa Lopez-Gil, Elena Vara
ABSTRACT
Lung resection surgery is associated with an inflammatory reaction. The use of 1-lung ventilation (OLV) seems to increase the likelihood of this reaction. Different prophylactic and therapeutic measures have been investigated to prevent lung injury secondary to OLV. Lidocaine, a commonly used local anesthetic drug, has antiinflammatory activity. Our main goal in this study was to investigate the effect of IV lidocaine on tumor necrosis factor α (TNF-α) lung expression during lung resection surgery with OLV. Eighteen pigs underwent left caudal lobectomy. The animals were divided into 3 groups: control, lidocaine, and sham. All animals received general anesthesia. In addition, animals in the lidocaine group received a continuous IV infusion of lidocaine during surgery (1.5 mg/kg/h). Animals in the sham group only underwent thoracotomy. Samples of bronchoalveolar lavage (BAL) fluid and plasma were collected before initiation of OLV, at the end of OLV, at the end of surgery, and 24 hours after surgery. Lung biopsy specimens were collected from the left caudal lobe (baseline) before surgery and from the mediastinal lobe and the left cranial lobe 24 hours after surgery. Samples were flash-frozen and stored to measure levels of the following inflammatory markers: interleukin (IL) 1β, IL-2, IL-10, TNF-α, nuclear factor κB, monocyte chemoattractant protein-1, inducible nitric oxide synthase, and endothelial nitric oxide synthase. Markers of apoptosis (caspase 3, caspase 9, Bad, Bax, and Bcl-2) were also measured. In addition, levels of metalloproteinases and nitric oxide metabolites were determined in BAL fluid and in plasma samples. A nonparametric test was used to examine statistical significance. OLV caused lung damage with increased TNF-α expression in BAL, plasma, and lung samples. Other inflammatory (IL-1β, nuclear factor κB, monocyte chemoattractant protein-1) and apoptosis (caspase 3, caspase 9, and BAX) markers were also increased. With the use of IV lidocaine there was a significant decrease in the levels of TNF-α in the same samples compared with the control group. Lidocaine administration also reduced the inflammatory and apoptotic changes observed in the control group. Hemodynamic values, blood gas values, and airway pressure were similar in all groups. Our results suggest that lidocaine can prevent OLV-induced lung injury through reduced expression of proinflammatory cytokines and lung apoptosis. Administration of lidocaine may help to prevent lung injury during lung surgery with OLV.
Product Description
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard
USP
Atracurium besylate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard