Merck
  • Transported substrate determines exchange rate in the multidrug resistance transporter EmrE.
Transported substrate determines exchange rate in the multidrug resistance transporter EmrE.
The Journal of biological chemistry (2014-01-23)
Emma A Morrison, Katherine A Henzler-Wildman
ABSTRACT
EmrE, a small multidrug resistance transporter, serves as an ideal model to study coupling between multidrug recognition and protein function. EmrE has a single small binding pocket that must accommodate the full range of diverse substrates recognized by this transporter. We have studied a series of tetrahedral compounds, as well as several planar substrates, to examine multidrug recognition and transport by EmrE. Here we show that even within this limited series, the rate of interconversion between the inward- and outward-facing states of EmrE varies over 3 orders of magnitude. Thus, the identity of the bound substrate controls the rate of this critical step in the transport process. The binding affinity also varies over a similar range and is correlated with substrate hydrophobicity within the tetrahedral substrate series. Substrate identity influences both the ground-state and transition-state energies for the conformational exchange process, highlighting the coupling between substrate binding and transport required for alternating access antiport.
Sigma-Aldrich
ISOGRO®-15N Powder -Growth Medium, 98 atom % 15N
Sigma-Aldrich
ISOGRO®-D Powder -Growth Medium, 97-99 atom % D
Sigma-Aldrich
ISOGRO®-15N,D Powder -Growth Medium, 98 atom % 15N, 97 atom % D
Sigma-Aldrich
ISOGRO®-13C,15N,D Powder -Growth Medium, 98 atom % 15N, 97-99 atom % D, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C Powder -Growth Medium, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C,15N Powder -Growth Medium, 98 atom % 15N, 99 atom % 13C