Merck
Performance evaluation of Samsung LABGEO(HC10) Hematology Analyzer.
Archives of pathology & laboratory medicine (2014-07-31)
Il Joong Park, Sunhyun Ahn, Young In Kim, Seon Joo Kang, Sung Ran Cho
ABSTRACT
The Samsung LABGEO(HC10) Hematology Analyzer (LABGEO(HC10)) is a recently developed automated hematology analyzer that uses impedance technologies. The analyzer provides 18 parameters including 3-part differential at a maximum rate of 80 samples per hour. To evaluate the performance of the LABGEO(HC10). We evaluated precision, linearity, carryover, and relationship for complete blood cell count parameters between the LABGEO(HC10) and the LH780 (Beckman Coulter Inc) in a university hospital in Korea according to the Clinical and Laboratory Standards Institute guidelines. Sample stability and differences due to the anticoagulant used (K₂EDTA versus K₃EDTA) were also evaluated. The LABGEO(HC10) showed linearity over a wide range and minimal carryover (<1%) for white blood cell, hemoglobin, red blood cell, and platelet parameters. Correlation between the LABGEO(HC10) and the LH780 was good for all complete blood cell count parameters (R > 0.92) except for mean corpuscular hemoglobin concentration. The bias estimated was acceptable for all parameters investigated except for monocyte count. Most parameters were stable until 24 hours both at room temperature and at 4°C. The difference by anticoagulant type was statistically insignificant for all parameters except for a few red cell parameters. The accurate results achievable and simplicity of operation make the unit recommendable for small to medium-sized laboratories.
Product Description
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Disodium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)