Merck
  • Automated derivatization and fluorimetric determination of biogenic amines in milk by zone fluidics coupled to liquid chromatography.
Automated derivatization and fluorimetric determination of biogenic amines in milk by zone fluidics coupled to liquid chromatography.
Journal of chromatography. A (2014-07-06)
Maria Notou, Anastasia Zotou, Paraskevas D Tzanavaras, Demetrius G Themelis
ABSTRACT
A novel zone-fluidics/high pressure liquid chromatographic (ZF-HPLC) method is described for the simultaneous determination of six biogenic monoamines, in the presence of hexylamine as internal standard. Automated on-line derivatization of the analytes with naphthalene-2,3-dicarboxaldehyde/cyanide ions was performed using the ZF concept and the derivatives were injected on the HPLC column for separation/detection. The influence of the ZF operation conditions on the derivatization reaction was investigated. The isoindoles formed were separated on a Kromasil C18 column (250 mm × 4 mm i.d., 5 μm), using an isocratic mobile phase of methanol/water (80:20, v/v) at a flow rate of 1.0 mL min(-1). Monitoring and quantification was carried out by fluorescence detection at 424/494 nm. The limits of detection were at the pg level with a sample volume of 20 μL. The whole procedure was evaluated and fully validated for the determination of biogenic amines in milk samples.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)