Merck
  • Pharmacologically and Edinger-Westphal stimulated accommodation in rhesus monkeys does not rely on changes in anterior chamber pressure.
Pharmacologically and Edinger-Westphal stimulated accommodation in rhesus monkeys does not rely on changes in anterior chamber pressure.
Experimental eye research (2014-07-02)
Lin He, Mark Wendt, Adrian Glasser
ABSTRACT
This study was undertaken to understand the role of anterior chamber pressure (ACP) during pharmacological and Edinger-Westphal (EW) stimulated accommodation in anesthetized monkeys. Experiments were performed on one iridectomized eye each of 7 anesthetized adolescent rhesus monkeys. Accommodation was induced by EW stimulation (n = 2) and intravenous administration of 0.25-4.0 mg/kg pilocarpine (n = 6). Accommodative refractive and biometric changes were measured with continuous 60 Hz infrared photorefraction (n = 6) and 100 Hz A-scan ultrasound biometry (n = 1). An ocular perfusion system was used to measure and manipulate ACP. Pressure was recorded via a 27-gauge needle in the anterior chamber connected to a pressure transducer (n = 7). The needle was also connected to a fluid reservoir to allow ACP to be manipulated and clamped (n = 4) by raising or lowering the fluid reservoir. In all six pharmacologically stimulated monkeys ACP increased during accommodation, from 0.70 to 2.38 mmHg, four of which showed pressure decreases preceding the pressure increases. Two eyes also showed increases in ACP during EW-stimulated accommodation of 2.8 and 7.2 mmHg. ACP increased with increasing EW stimulus amplitudes (n = 2). Clamping or externally manipulating ACP had no effect on resting refraction or on EW and pharmacologically stimulated accommodation in four eyes. The results show that EW stimulated and pharmacologically stimulated accommodation do not rely on ACP in rhesus monkeys.
Product Description
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Pilocarpine nitrate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications
Pilocarpine nitrate, European Pharmacopoeia (EP) Reference Standard