Merck
  • A novel straightforward synthesis of 2,4-disubstituted-1,3,5-triazines via aerobic copper-catalyzed cyclization of amidines with DMF.
A novel straightforward synthesis of 2,4-disubstituted-1,3,5-triazines via aerobic copper-catalyzed cyclization of amidines with DMF.
Organic letters (2014-06-17)
Xiaowen Xu, Min Zhang, Huanfeng Jiang, Jia Zheng, Yiqun Li
ABSTRACT
A novel straightforward synthesis of both symmetrical and unsymmetrical 2,4-disubstituted-1,3,5-triazines via aerobic copper-catalyzed cyclization of amidines with DMF as a one-carbon synthon has been developed. The presented method allows synthesizing the products that are currently inaccessible or challenging to prepare with the advantages of operational simplicity, broad substrate scope, and no need for prefunctionalized reagents, making it a highly practical approach to access various 2,4-disubstituted-1,3,5-triazines.
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Copper, insulated wire, 50m, conductor diameter 0.24mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, insulated wire, 50m, conductor diameter 0.45mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, microfoil, disks, 25mm, thinness 0.25μm, specific density 219.9μg/cm2, permanent mylar 3.5μm support, 99.99+%
Copper - O.F.H.C., foil, 50x50mm, thickness 2.0mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 300x300mm, thickness 0.20mm, hard, 99.95+%
Copper, insulated wire, 5m, conductor diameter 0.1mm, insulation thickness 0.015mm, 10-30 micron polyesterimide insulation, 99.9%
Copper, insulated wire, 2m, conductor diameter 1.0mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper - O.F.H.C., foil, 0.5m coil, thickness 0.006mm, hard, 99.95+%
Copper - O.F.H.C., foil, 150x150mm, thickness 2.0mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 300x300mm, thickness 0.25mm, half hard, 99.95+%
Copper, microfoil, disks, 10mm, thinness 0.25μm, specific density 219.9μg/cm2, permanent mylar 3.5μm support, 99.99+%
Copper - O.F.H.C., foil, 50x50mm, thickness 0.1mm, hard, 99.95+%
Copper, insulated wire, 50m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Copper, insulated wire, 20m, conductor diameter 0.018mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%