Merck
  • Imaging insights into basal ganglia function, Parkinson's disease, and dystonia.
Imaging insights into basal ganglia function, Parkinson's disease, and dystonia.
Lancet (London, England) (2014-06-24)
A Jon Stoessl, Stephane Lehericy, Antonio P Strafella
ABSTRACT
Recent advances in structural and functional imaging have greatly improved our ability to assess normal functions of the basal ganglia, diagnose parkinsonian syndromes, understand the pathophysiology of parkinsonism and other movement disorders, and detect and monitor disease progression. Radionuclide imaging is the best way to detect and monitor dopamine deficiency, and will probably continue to be the best biomarker for assessment of the effects of disease-modifying therapies. However, advances in magnetic resonance enable the separation of patients with Parkinson's disease from healthy controls, and show great promise for differentiation between Parkinson's disease and other akinetic-rigid syndromes. Radionuclide imaging is useful to show the dopaminergic basis for both motor and behavioural complications of Parkinson's disease and its treatment, and alterations in non-dopaminergic systems. Both PET and MRI can be used to study patterns of functional connectivity in the brain, which is disrupted in Parkinson's disease and in association with its complications, and in other basal-ganglia disorders such as dystonia, in which an anatomical substrate is not otherwise apparent. Functional imaging is increasingly used to assess underlying pathological processes such as neuroinflammation and abnormal protein deposition. This imaging is another promising approach to assess the effects of treatments designed to slow disease progression.
Product Description
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dopamine hydrochloride
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®