Merck
  • EDTA chelation therapy alone and in combination with oral high-dose multivitamins and minerals for coronary disease: The factorial group results of the Trial to Assess Chelation Therapy.
EDTA chelation therapy alone and in combination with oral high-dose multivitamins and minerals for coronary disease: The factorial group results of the Trial to Assess Chelation Therapy.
American heart journal (2014-06-24)
Gervasio A Lamas, Robin Boineau, Christine Goertz, Daniel B Mark, Yves Rosenberg, Mario Stylianou, Theodore Rozema, Richard L Nahin, L Terry Chappell, Lauren Lindblad, Eldrin F Lewis, Jeanne Drisko, Kerry L Lee
ABSTRACT
Disodium ethylenediaminetetraacetic acid (EDTA) reduced adverse cardiac outcomes in a factorial trial also testing oral vitamins. This report describes the intent-to-treat comparison of the 4 factorial groups overall and in patients with diabetes. This was a double-blind, placebo-controlled, 2 × 2 factorial multicenter randomized trial of 1,708 post-myocardial infarction (MI) patients ≥50 years of age and with creatinine ≤2.0 mg/dL randomized to receive 40 EDTA chelation or placebo infusions plus 6 caplets daily of a 28-component multivitamin-multimineral mixture or placebo. The primary end point was a composite of total mortality, MI, stroke, coronary revascularization, or hospitalization for angina. Median age was 65 years, 18% were female, 94% were Caucasian, 37% were diabetic, 83% had prior coronary revascularization, and 73% were on statins. Five-year Kaplan-Meier estimates for the primary end point was 31.9% in the chelation + high-dose vitamin group, 33.7% in the chelation + placebo vitamin group, 36.6% in the placebo infusion + active vitamin group, and 40.2% in the placebo infusions + placebo vitamin group. The reduction in primary end point by double active treatment compared with double placebo was significant (hazard ratio 0.74, 95% CI 0.57-0.95, P = .016). In patients with diabetes, the primary end point reduction of double active compared with double placebo was more pronounced (hazard ratio 0.49, 95% CI 0.33-0.75, P < .001). In stable post-MI patients on evidence-based medical therapy, the combination of oral high-dose vitamins and chelation therapy compared with double placebo reduced clinically important cardiovascular events to an extent that was both statistically significant and of potential clinical relevance.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Disodium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)