Merck
  • Gestational and lactational exposure to the polychlorinated biphenyl mixture Aroclor 1254 modulates retinoid homeostasis in rat offspring.
Gestational and lactational exposure to the polychlorinated biphenyl mixture Aroclor 1254 modulates retinoid homeostasis in rat offspring.
Toxicology letters (2014-06-03)
Javier Esteban, Lubna E Elabbas, Daniel Borg, Maria Herlin, Agneta Åkesson, Xavier Barber, Gerd Hamscher, Heinz Nau, Wayne J Bowers, Jamie S Nakai, Matti Viluksela, Helen Håkansson
ABSTRACT
Polychlorinated biphenyls (PCBs) induce a broad spectrum of biochemical and toxic effects in mammals including alterations of the vital retinoid (vitamin A) system. The aim of this study was to characterize alterations of tissue retinoid levels in rat offspring and their dams following gestational and lactational exposure to the PCB mixture Aroclor 1254 (A1254) and to assess the interrelationship of these changes with other established sensitive biochemical and toxicological endpoints. Sprague-Dawley rat dams were exposed orally to 0 or 15 mg/kg body weight/day of A1254 from gestational day 1 to postnatal day (PND) 23. Livers, kidneys and serum were collected from the offspring on PNDs 35, 77 and 350. Tissue and serum retinoid levels, hepatic cytochrome P450 (CYP) enzymes and serum thyroid hormones were analyzed. A multivariate regression between A1254 treatment, hepatic retinoid levels, hepatic CYP enzymes activities, thyroid hormone levels and body/liver weights was performed using an orthogonal partial least-squares (PLS) analysis. The contribution of dioxin-like (DL) components of A1254 to the observed effects was also estimated using the toxic equivalency (TEQ) concept. In both male and female offspring short-term alterations in tissue retinoid levels occurred at PND35, i.e. decreased levels of hepatic retinol and retinoic acid (RA) metabolite 9-cis-4-oxo-13,14-dihydro-RA with concurrent increases in hepatic and renal all-trans-RA levels. Long-term changes consisted of decreased hepatic retinyl palmitate and increased renal retinol levels that were apparent until PND350. Retinoid system alterations were associated with altered CYP enzyme activities and serum thyroid hormone levels as well as body and liver weights in both offspring and dams. The estimated DL activity was within an order of magnitude of the theoretical TEQ for different endpoints, indicating significant involvement of DL congeners in the observed effects. This study shows that tissue retinoid levels are affected both short- and long-term by developmental A1254 exposure and are associated with alterations of other established endpoints of toxicological concern.
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254 solution, certified reference material, 1000 μg/mL in isooctane
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®