Merck
  • Marginally excessive iron loading transiently blocks mucosal iron uptake in iron-deficient rats.
Marginally excessive iron loading transiently blocks mucosal iron uptake in iron-deficient rats.
American journal of physiology. Gastrointestinal and liver physiology (2014-05-17)
Shoko Shinoda, Shiho Yoshizawa, Eriko Nozaki, Kouki Tadai, Anna Arita
ABSTRACT
Regular "mucosal block" is characterized by decreased uptake of a normal iron load 3-72 h after the administration of excess iron (generally 10 mg) to iron-deficient animals. We found that short-acting mucosal block could be induced by much lower iron concentration and much shorter induction time than previously reported, without affecting levels of gene expression. A rapid endocytic mechanism was reported to decrease intestinal iron absorption after a high iron load, but the activating iron load and the time to decreased absorption were undetermined. We assessed the effects of 30-2,000 μg iron load on iron uptake in the duodenal loop of iron-deficient and iron-sufficient rats under anesthesia. One hour later, mucosal cellular iron uptake in iron-deficient rats administered 30 μg iron was 76.1%, decreasing 25% to 50.7% in rats administered 2,000 μg iron. In contrast, iron uptake by iron-sufficient rats was 63% (range 60.3-65.5%) regardless of iron load. Duodenal mucosal iron concentration was significantly lower in iron-deficient than in iron-sufficient rats. Iron levels in portal blood were consistently higher in iron-deficient rats regardless of iron load, in contrast to the decreased iron uptake on the luminal side. Iron loading blocked mucosal uptake of marginally excess iron (1,000 μg), with a greater effect at 15 min than at 30 min. The rapid induction of short-acting mucosal block only in iron-deficient rats suggests DMT1 internalization.
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL