Merck
  • Use of Fluorinated Compounds To Detect Aromatic Metabolites from m-Cresol in a Methanogenic Consortium: Evidence for a Demethylation Reaction.
Use of Fluorinated Compounds To Detect Aromatic Metabolites from m-Cresol in a Methanogenic Consortium: Evidence for a Demethylation Reaction.
Applied and environmental microbiology (1993-07-01)
K L Londry, P M Fedorak
ABSTRACT
Anaerobic sewage sludge was used to enrich a methanogenic m-cresol-degrading consortium. 6-Fluoro-3-methylphenol was synthesized and added to subcultures of the consortium with m-cresol. This caused the accumulation of 4-hydroxy-2-methylbenzoic acid. In a separate experiment, the addition of 3-fluorobenzoic acid caused the transient accumulation of 4-hydroxybenzoic acid. Inhibition with bromoethanesulfonic acid caused the accumulation of benzoic acid. Thus, the proposed degradation pathway was m-cresol --> 4-hydroxy-2-methylbenzoic acid --> 4-hydroxybenzoic acid --> benzoic acid. The m-cresol-degrading consortium was able to convert exogenous 4-hydroxybenzoic acid and benzoic acid to methane. In addition, for each metabolite of m-cresol identified, the corresponding fluorinated metabolite was detected, giving the following sequence: 6-fluoro-3-methylphenol --> 5-fluoro-4-hydroxy-2-methylbenzoic acid --> 3-fluoro-4-hydroxybenzoic acid --> 3-fluorobenzoic acid. The second step in each of these pathways is a novel demethylation which was rate limiting. This demethylation reaction would likely facilitate the transformation of the methyl group to methane, which is consistent with the results of a previous study that showed that the methyl carbon of m-[methyl-C]cresol was recovered predominantly as [C]methane (D. J. Roberts, P. M. Fedorak, and S. E. Hrudey, Can. J. Microbiol. 33:335-338, 1987). The final aromatic compound in the proposed route for m-cresol metabolism was benzoic acid, and its detection in these cultures merges the pathway for the methanogenic degradation of m-cresol with those for the anaerobic metabolism of many phenols.
Sigma-Aldrich
4-Fluoro-3-methylphenol, 98%
Pricing and availability is not currently available.