Merck
Atenolol-induced lupus erythematosus.
Journal of the American Academy of Dermatology (1997-08-01)
M McGuiness, R A Frye, J S Deng
ABSTRACT
Atenolol is a beta-blocker commonly used for treating hypertension. It can induce various kinds of adverse side effects, including psoriasiform skin eruptions, skin necrosis, vasculitis, and (rarely) drug-induced connective tissue disease. We encountered a patient receiving atenolol for his hypertension for 3 years who subsequently acquired connective tissue disease and antihistone antibodies. The initial serologic antinuclear antibody test was negative at a dilution of 1/20 but was positive after further serial dilutions, indicating the prozone phenomenon as the cause of the false-negative result. Six months after discontinuation of atenolol, the skin rash disappeared and antihistone antibody subsided. His skin rash reappeared on rechallenge with atenolol for 3 days, confirming that atenolol was responsible for his lupus erythematosus.
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Atenolol, European Pharmacopoeia (EP) Reference Standard
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard