Merck
Occupational phosgene poisoning: a case report and review.
Journal of accident & emergency medicine (1995-09-01)
J P Wyatt, C A Allister
ABSTRACT
Phosgene is a highly toxic gas to which some workers may be occupationally exposed. This case report demonstrates the possibility of refrigeration workers suffering phosgene poisoning after heating certain chlorinated fluorocarbons ('freons'). The need to suspect phosgene exposure and observe such patients is emphasized, especially in view of the delay in clinical deterioration observed in some patients who subsequently develop adult respiratory distress syndrome.
Sigma-Aldrich
Phosgene solution, ~20% in toluene
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene