- Chymotrypsin: molecular and catalytic properties.
Chymotrypsin: molecular and catalytic properties.
Clinical biochemistry (1986-12-01)
W Appel
PMID3555886
ABSTRACT
A review on the peptide hydrolase chymotrypsin gives some characteristics concerning the structure, specificity, mechanism of activation from its precursor and catalytic activity. Problems of the active sites, allosteric alterations, changes of conformation, different behaviour against types of substrates, and suggested kinetics are shown in some detail. Finally, the diagnostic role of this serine proteinase is mentioned.
Product Description
Chymotrypsin, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder