Merck
Cerium oxide nanoparticles: applications and prospects in nanomedicine.
Nanomedicine (London, England) (2013-08-31)
Soumen Das, Janet M Dowding, Kathryn E Klump, James F McGinnis, William Self, Sudipta Seal
ABSTRACT
Promising results have been obtained using cerium (Ce) oxide nanoparticles (CNPs) as antioxidants in biological systems. CNPs have unique regenerative properties owing to their low reduction potential and the coexistence of both Ce(3+)/Ce(4+) on their surfaces. Defects in the crystal lattice due to the presence of Ce(3+) play an important role in tuning the redox activity of CNPs. The surface Ce(3+):Ce(4+) ratio is influenced by the microenvironment. Therefore, the microenvironment and synthesis method adopted also plays an important role in determining the biological activity and toxicity of CNPs. The presence of a mixed valance state plays an important role in scavenging reactive oxygen and nitrogen species. CNPs are found to be effective against pathologies associated with chronic oxidative stress and inflammation. CNPs are well tolerated in both in vitro and in vivo biological models, which makes CNPs well suited for applications in nanobiology and regenerative medicine.
Product Description
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips