Merck
Mitomycin C in the treatment of keloids: a case and review.
Journal of drugs in dermatology : JDD (2013-07-11)
Rebecca Kleinerman, Suzanne L Kilmer, Vera A Chotzen
ABSTRACT
Mitomycin C (MMC) is an antineoplastic antibiotic that has been used off-label in the treatment of hypertrophic scars and keloids. Herein we report our successful use of this agent in a patient with sternal keloids refractory to other means of therapy. We further review the literature regarding the use of MMC in the treatment of keloids.
Product Description
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture