Merck
  • Behavior and impact of zirconium in the soil-plant system: plant uptake and phytotoxicity.
Behavior and impact of zirconium in the soil-plant system: plant uptake and phytotoxicity.
Reviews of environmental contamination and toxicology (2012-10-24)
Muhammad Shahid, Emmanuel Ferrand, Eva Schreck, Camille Dumat
ABSTRACT
Zirconium (Zr) is a transition metal that has both stable and radioactive isotopes.This metal has gained significant attention as a major pollutant of concern, partly because it has been prominent in the debate concerning the growing anthropogenic pressure on the environment. Its numerous past and present uses have induced significant soil and water pollution. Zr is generally considered to have low mobility in soils. The behavior of Zr particularly depends on the characteristics of the media in which it exists, and even its presence in the biosphere as a contaminate may affect its behavior. In this chapter, we describe the relationship between the behavior of Zrand its speciation in soils, its uptake and accumulation by plants, its translocation and toxicity inside plants, and mechanisms by which plants detoxify it.Zr is abundant and occurs naturally in the earth's crust. Zr emissions to the atmosphere are increasing from anthropogenic activities such as its use in industry and nuclear reactors. Zr forms various complexes with soil components, which reduces its soil mobility and phytoavailabilty. The mobility and phytoavailabilty of Zr in soil depend on its speciation and the physicochemical properties of soil that include soil pH, texture, and organic contents. Despite having low soil mobility and phytoavailability,amounts of Zr are absorbed by plants, mainly through the root system and can thereby enter the food chain.After plant uptake, Zr mainly accumulates in root cells. Zr does not have any known essential function in plant or animal metabolism. Although little published data are available, we conclude that the phytotoxicity of Zr is generally low.Notwithstanding, Zr can significantly reduce plant growth and can affect plantenzyme activity. When exposed to Zr-induced toxicity, plants possess numerous defense mechanisms to cope with the toxicity. Such strategies include Zr sequestration in plant roots and activation of various antioxidants. Because Zr may have impact on the biosphere, we believe it deserves to be evaluated in supplementary studies that will enhance the understanding of its behavior in soil-plant systems.
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Zirconium, foil, light tested, 100x100mm, thickness 0.015mm, 99.2%
Zirconium, rod, 100mm, diameter 3.0mm, centerless ground, 99.2%
Zirconium, rod, 100mm, diameter 5.0mm, centerless ground, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.015mm, 99.2%
Zirconium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.2%
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 300x300mm, thickness 0.125mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.016mm, 99.2%
Zirconium, foil, 1m coil, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zirconium, foil, 10m coil, thickness 0.02mm, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.007mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.007mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.02mm, 99.2%
Zirconium, foil, light tested, 100x300mm, thickness 0.016mm, 99.2%
Zirconium, foil, not light tested, 100x100mm, thickness 0.003mm, 99.2%
Zirconium, foil, light tested, 150x150mm, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.008mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.0125mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%