Merck
High-valent organometallic copper and palladium in catalysis.
Nature (2012-04-14)
Amanda J Hickman, Melanie S Sanford
ABSTRACT
Copper and palladium catalysts are critically important in numerous commercial chemical processes. Improvements in the activity, selectivity and scope of these catalysts could drastically reduce the environmental impact, and increase the sustainability, of chemical reactions. One rapidly developing strategy for achieving these goals is to use 'high-valent' organometallic copper and palladium intermediates in catalysis. Here we describe recent advances involving both the fundamental chemistry and the applications of these high-valent metal complexes in numerous synthetically useful catalytic transformations.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Palladium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, wire reel, 1m, diameter 0.125mm, hard, 99.95%
Palladium, wire reel, 0.1m, diameter 0.25mm, hard, 99.95%
Palladium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.95%
Palladium, tube, 120mm, outside diameter 6mm, inside diameter 5mm, wall thickness 0.5mm, as drawn, 99.95%
Palladium, wire reel, 10m, diameter 0.025mm, as drawn, 99.9%
Palladium, wire reel, 1m, diameter 0.25mm, as drawn, 99.99+%
Palladium, wire reel, 0.05m, diameter 1.0mm, hard, 99.95%
Palladium, wire reel, 0.2m, diameter 1.0mm, hard, 99.95%
Palladium, wire reel, 2m, diameter 0.025mm, as drawn, 99.9%
Palladium, wire reel, 5m, diameter 0.05mm, as drawn, 99.9%
Palladium, wire reel, 20m, diameter 0.025mm, as drawn, 99.9%
Palladium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.99+%
Palladium, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, wire reel, 25m, diameter 0.125mm, hard, 99.95%