Merck
  • Conversion of linoleic acid and alpha-linolenic acid to long-chain polyunsaturated fatty acids (LCPUFAs), with a focus on pregnancy, lactation and the first 2 years of life.
Conversion of linoleic acid and alpha-linolenic acid to long-chain polyunsaturated fatty acids (LCPUFAs), with a focus on pregnancy, lactation and the first 2 years of life.
Maternal & child nutrition (2011-03-05)
Robert A Gibson, Bev Muhlhausler, Maria Makrides
ABSTRACT
Over the past two decades, there has been a marked shift in the fatty acid composition of the diets of industrialized nations towards increased intake of the n-6 fatty acid linoleic acid (LA, 18:2n-6), largely as a result of the replacement of saturated fats with plant-based polyunsaturated fatty acid (PUFA). While health agencies internationally continue to advocate for high n-6 PUFA intake combined with increased intakes of preformed n-3 long-chain PUFAs (LCPUFA) docosahexaenoic acid (DHA, 22:6n-3) and eicosapentaenoic acid (EPA, 20:5n-3) to reduce the incidence of cardiovascular disease (CVD), there are questions as to whether this is the best approach. LA competes with alpha-linolenic acid (18:3n-3) for endogenous conversion to the LC derivatives EPA and DHA, and LA also inhibits incorporation of DHA and EPA into tissues. Thus, high-LA levels in the diet generally result in low n-3 LCPUFA status. Pregnancy and infancy are developmental periods during which the fatty acid supply is particularly critical. The importance of an adequate supply of n-3 LCPUFA for ensuring optimal development of infant brain and visual systems is well established, and there is now evidence that the supply of n-3 LCPUFA also influences a range of growth, metabolic and immune outcomes in childhood. This review will re-evaluate the health benefits of modern Western diets and pose the question of whether the introduction of similar diets to nations with emerging economies is the most prudent public health strategy for improving health in these populations.
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Linolenic acid, analytical standard
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture