Merck
  • Olmesartan medoxomil, amlodipine besylate and hydrochlorothiazide triple combination for hypertension.
Olmesartan medoxomil, amlodipine besylate and hydrochlorothiazide triple combination for hypertension.
Expert review of cardiovascular therapy (2010-12-21)
C Venkata S Ram
ABSTRACT
Hypertension is an increasingly prevalent cardiovascular risk factor associated with high rates of morbidity and mortality. Lowering blood pressure (BP) to recommended levels reduces the risk of hypertension-associated cardiovascular events. Despite current treatment options and recommendations, the BP of many patients remains suboptimally controlled. There is a need for more effective management in patients not achieving BP goals on current monotherapy or combination therapy. Regimens combining three or more antihypertensive agents have been shown to increase the proportion of patients achieving BP goals. Recent data suggest that the combination of olmesartan medoxomil, amlodipine and hydrochlorothiazide is a rational treatment option offering safe and effective BP reductions and goal attainment in a greater proportion of patients compared with dual-combination regimens. As the understanding of the importance of BP control grows, treatment options that enable patients to achieve BP goals quickly and safely will become increasingly important in hypertension management.
Product Description
Amlodipine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Amlodipine besylate, United States Pharmacopeia (USP) Reference Standard
Amlodipine besylate, European Pharmacopoeia (EP) Reference Standard
Supelco
Amlodipine besylate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)