Merck
  • Final report of the safety assessment of allantoin and its related complexes.
Final report of the safety assessment of allantoin and its related complexes.
International journal of toxicology (2010-05-22)
Lillian C Becker, Wilma F Bergfeld, Donald V Belsito, Curtis D Klaassen, James G Marks, Ronald C Shank, Thomas J Slaga, Paul W Snyder, F Alan Andersen
ABSTRACT
Allantoin is a heterocyclic organic compound. Allantoin ascorbate, allantoin biotin, allantoin galacturonic acid, allantoin glycyrrhetinic acid, allantoin panthenol, and allantoin polygalacturonic acid are complexes of allantoin. All of the ingredients in this review act as skin-conditioning agents. Allantoin was reported to be used in 1376 cosmetic products at concentrations up to 2%. There are data gaps regarding use and concentration of the remaining allantoin complexes. Ascorbic acid, biotin, glycyrrhetinic acid, and panthenol have been determined by the CIR Expert Panel to be safe. Galacturonic acid and polygalacturonic acid have not been reviewed by the CIR Expert Panel, and substantial data on these chemicals were not available. The safety test data in this safety assessment and in previous safety assessments were considered sufficient to support the safety of allantoin and the allantoin complexes in product categories and at concentrations reviewed in this safety assessment.
Product Description
Allantoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Allantoin, analytical standard
Supelco
Allantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Allantoin, ≥98.0% (N)