Merck
Neurotoxicity of toluene.
Toxicology letters (2010-07-06)
Tin-Tin Win-Shwe, Hidekazu Fujimaki
ABSTRACT
Chemical susceptibility is triggered by a large range of chemicals present both indoors and outdoors including pesticides, cleaning products, perfumes, scented products and cigarette smoke. Health risk after chemical exposure depends on age, sex, genetic factors, socioeconomic status, nutritional status, and environmental factors. Toluene is one of volatile organic chemicals that causes different sensitivity in individuals. Although neurotoxic and immunotoxic effects of toluene have been studied extensively, the underlying mechanism remains obscure. This review highlights the possible neuroimmune factors influencing toluene sensitivity and neurotoxicity in a mouse model.
Product Description
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%