Merck
  • Fermentation of lactose to bio-ethanol by yeasts as part of integrated solutions for the valorisation of cheese whey.
Fermentation of lactose to bio-ethanol by yeasts as part of integrated solutions for the valorisation of cheese whey.
Biotechnology advances (2010-02-16)
Pedro M R Guimarães, José A Teixeira, Lucília Domingues
ABSTRACT
Cheese whey, the main dairy by-product, is increasingly recognized as a source of many bioactive valuable compounds. Nevertheless, the most abundant component in whey is lactose (ca. 5% w/v), which represents a significant environmental problem. Due to the large lactose surplus generated, its conversion to bio-ethanol has long been considered as a possible solution for whey bioremediation. In this review, fermentation of lactose to ethanol is discussed, focusing on wild lactose-fermenting yeasts, particularly Kluyveromyces marxianus, and recombinant Saccharomyces cerevisiae strains. The early efforts in the screening and characterization of the fermentation properties of wild lactose-consuming yeasts are reviewed. Furthermore, emphasis is given on the latter advances in engineering S. cerevisiae strains for efficient whey-to-ethanol bioprocesses. Examples of industrial implementation are briefly discussed, illustrating the viability of whey-to-ethanol systems. Current developments on strain engineering together with the growing market for biofuels will likely boost the industrial interest in such processes.
Product Description
USP
Anhydrous lactose, United States Pharmacopeia (USP) Reference Standard
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard