Merck
  • Sodium bicarbonate for the prevention of contrast induced-acute kidney injury: a systematic review and meta-analysis.
Sodium bicarbonate for the prevention of contrast induced-acute kidney injury: a systematic review and meta-analysis.
Clinical journal of the American Society of Nephrology : CJASN (2009-08-29)
Somjot S Brar, Swapnil Hiremath, George Dangas, Roxana Mehran, Simerjeet K Brar, Martin B Leon
ABSTRACT
Infusion of sodium bicarbonate has been suggested as a preventative strategy but reports are conflicting on its efficacy. The aim of this study was to assess the effectiveness of hydration with sodium bicarbonate for the prevention of contrast-induced acute kidney injury (CI-AKI). Medline, EMBASE, Cochrane library, and the Internet were searched for randomized controlled trials comparing hydration between sodium bicarbonate and chloride for the prevention of CI-AKI between 1966 and November 2008. Fourteen trials that included 2290 patients were identified. There was significant heterogeneity between studies (P heterogeneity = 0.02; I(2) = 47.8%), which was largely accounted for by trial size (P = 0.016). Trials were therefore classified by size. Three trials were categorized as large (n = 1145) and 12 as small (n = 1145). Among the large trials, the incidence of CI-AKI for sodium bicarbonate and sodium chloride was 10.7 and 12.5%, respectively; the relative risk (RR) [95% confidence interval (CI)] was 0.85 (0.63 to 1.16) without evidence of heterogeneity (P = 0.89, I(2) = 0%). The pooled RR (95% CI) among the 12 small trials was 0.50 (0.27 to 0.93) with significant between-trial heterogeneity (P = 0.01; I(2) = 56%). The small trials were more likely to be of lower methodological quality. A significant clinical and statistical heterogeneity was observed that was largely explained by trial size and published status. Among the large randomized trials there was no evidence of benefit for hydration with sodium bicarbonate compared with sodium chloride for the prevention of CI-AKI. The benefit of sodium bicarbonate was limited to small trials of lower methodological quality.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C