Merck
  • A comprehensive review of the loop diuretics: should furosemide be first line?
A comprehensive review of the loop diuretics: should furosemide be first line?
The Annals of pharmacotherapy (2009-10-22)
Kurt A Wargo, William M Banta
ABSTRACT
To review the literature regarding the pharmacokinetic profiles, comparative safety and efficacy, and comparative costs of loop diuretics to evaluate the current clinical usefulness of furosemide. A search of MEDLINE (1966-June 2009) was conducted using the terms furosemide, torsemide, bumetanide, ethacrynic acid, and loop diuretics. Articles were limited to those written in English. All English-language articles identified from the data sources were reviewed. Studies were eligible if they encompassed pharmacokinetics, comparative safety and efficacy, or comparative costs of the loop diuretics. In patients with heart failure (HF), torsemide demonstrated decreased mortality compared with furosemide in 1 study (2.2% vs 4.5% in the furosemide group; p < 0.05), decreased hospitalizations in 1 study (23 in the torsemide group vs 61 in the furosemide group; p < 0.01), and improved New York Heart Association functional classifications in 2 studies. In the first, 45.8% with torsemide versus 37.2% with furosemide demonstrated improvement in at least one functional class (p = 0.00017). In the second, 40.2% with torsemide and 30.7% with furosemide demonstrated improvement in at least one functional class (p = 0.014). In 2 of 3 studies of patients with cirrhosis, torsemide increased natriuresis and total volume diuresed compared with furosemide in patients with cirrhosis; however, no significant difference between the agents with respect to plasma renin and aldosterone concentrations was demonstrated. In patients with pulmonary hypertension, central venous pressure, capillary wedge pressure, and stroke volume significantly improved from baseline among patients who received torsemide, but not in those who received furosemide, although the intergroup analysis failed to reach statistical significance. Among patients with chronic kidney disease, no significant differences were noted with respect to natriuresis and blood pressure control between the 2 agents; however, in patients with acute kidney injury, patients who received furosemide had a significant improvement in urine output versus the torsemide group. Additionally, 2 trials comparing bumetanide with furosemide were identified, although the results were conflicting. In patients with nephrotic syndrome, bumetanide significantly improved weight loss in the first 4 weeks and in week 20, compared with furosemide. In patients with HF, significant improvement in dyspnea at rest and on exertion was exhibited in the bumetanide group, but not in the furosemide group; no significant difference was noted between the 2 groups when evaluating global assessment. Growing evidence demonstrates more favorable pharmacokinetic profiles of torsemide and bumetanide compared with furosemide. Furthermore, torsemide may be more efficacious and safer than furosemide in patients with HF. A trial comparing all 3 drugs would be required to confirm torsemide as the primary loop diuretic in patients with HF, but based upon limited current evidence, we recommend torsemide over furosemide. Currently, little evidence exists to support either torsemide or bumetanide as first-line treatment over furosemide in patients with other edematous disease states.
Product Description
Furosemide for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Furosemide
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®