Merck
PSGL-1 function in immunity and steady state homeostasis.
Immunological reviews (2009-07-15)
Douglas A Carlow, Klaus Gossens, Silvia Naus, Krystle M Veerman, Wooseok Seo, Hermann J Ziltener
ABSTRACT
The substantial importance of P-selectin glycoprotein ligand 1 (PSGL-1) in leukocyte trafficking has continued to emerge beyond its initial identification as a selectin ligand. PSGL-1 seemed to be a relatively simple molecule with an extracellular mucin domain extended as a flexible rod, teleologically consistent with its primary role in tethering leukocytes to endothelial selectins. The rolling interaction between leukocyte and endothelium mediated by this selectin-PSGL-1 interaction requires branched O-glycan extensions on specific PSGL-1 amino acid residues. In some cells, such as neutrophils, the glycosyltransferases involved in formation of the O-glycans are constitutively expressed, while in other cells, such as T cells, they are expressed only after appropriate activation. Thus, PSGL-1 supports leukocyte recruitment in both innate and adaptive arms of the immune response. A complex array of amino acids within the selectins engage multiple sugar residues of the branched O-glycans on PSGL-1 and provide the molecular interactions responsible for the velcro-like catch bonds that support leukocyte rolling. Such binding of PSGL-1 can also induce signaling events that influence cell phenotype and function. Scrutiny of PSGL-1 has revealed a better understanding of how it performs as a selectin ligand and yielded unexpected insights that extend its scope from supporting leukocyte rolling in inflammatory settings to homeostasis including stem cell homing to the thymus and mature T-cell homing to secondary lymphoid organs. PSGL-1 has been found to bind homeostatic chemokines CCL19 and CCL21 and to support the chemotactic response to these chemokines. Surprisingly, the O-glycan modifications of PSGL-1 that support rolling mediated by selectins in inflammatory conditions interfere with PSGL-1 binding to homeostatic chemokines and thereby limit responsiveness to the chemotactic cues used in steady state T-cell traffic. The multi-level influence of PSGL-1 on cell traffic in both inflammatory and steady state settings is therefore substantially determined by the orchestrated addition of O-glycans. However, central as specific O-glycosylation is to PSGL-1 function, in vivo regulation of PSGL-1 glycosylation in T cells remains poorly understood. It is our purpose herein to review what is known, and not known, of PSGL-1 glycosylation and to update understanding of PSGL-1 functional scope.