Merck
  • Polyvinylpyrrolidone-sodium hyaluronate gel (Gelclair): a bioadherent oral gel for the treatment of oral mucositis and other painful oral lesions.
Polyvinylpyrrolidone-sodium hyaluronate gel (Gelclair): a bioadherent oral gel for the treatment of oral mucositis and other painful oral lesions.
Expert opinion on drug metabolism & toxicology (2008-10-28)
Patricia C Buchsel
ABSTRACT
Painful oral lesions in patients with cancer, HIV or aphthous ulcers can cause significant pain and discomfort. Those with oral mucositis (OM) resulting from high-dose chemotherapy or irradiation are the most at risk for painful lesions that lead to significant morbidity and mortality, decreased quality of life and increased economic costs. Numerous approaches have been taken to prevent and treat OM in the cancer patient but this condition remains a significant obstacle to cancer therapy. Small studies have been done in healing oral lesions arising from laser surgery and in wound healing but research is needed to support the use of polyvinylpyrrolidone-sodium hyaluronate gel (Gelclair) in these clinical setting. The purpose of this paper is to review the benefit of polyvinylpyrrolidone-sodium hyaluronate gel, a topical oral mucosal barrier gel to prevent or diminish the pain of OM and other oral lesions. Primary resources in peer-reviewed journals, chapters in recognized textbooks and information from international and national oncology and dental medicine scientific meetings. Oncology patients receiving high-dose immunosuppressive therapy are the most at risk with painful OM. Current prevention and treatment options for OM have limited effectiveness. Although opioids are available for pain management, novel agents, to be used alone or in combination with other proven measures to decrease the severity and duration of OM, are emerging. Clinical studies in the use of polyvinylpyrrolidone-sodium hyaluronate to reduce the pain or disruption to the oral mucosa has merit. Few clinical trials or main publications have supported the use of this product. Robust randomized trials are needed to further support the clinical utility of this agent.
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Kollidon® 25
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Povidone, European Pharmacopoeia (EP) Reference Standard
USP
Crospovidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Povidone, United States Pharmacopeia (USP) Reference Standard
Crospovidone, European Pharmacopoeia (EP) Reference Standard