Merck
Lanthanum carbonate--a first line phosphate binder?
Seminars in dialysis (2007-07-20)
Declan de Freitas, Rosemary L Donne, Alastair J Hutchison
ABSTRACT
Hyperphosphatemia is associated with increased morbidity and mortality in dialysis patients. Oral phosphate binders are necessary to control serum phosphate in patients eating a normal diet and undergoing peritoneal dialysis or thrice weekly hemodialysis. Until recently, none of the available drugs came close to the model of an ideal oral binder, which would demonstrate high affinity, rapid binding regardless of pH, low solubility and absorption, lack of toxicity, palatability, and reasonable cost. Lanthanum carbonate is a safe and effective binder with data demonstrating no toxic effects after continuous exposure up to 6 years. Suggestions that absorption and accumulation of lanthanum is significant and of clinical importance do not stand up to close scrutiny and recommended it as a first line treatment for dialysis patients, particularly if they have evidence of vascular calcification.
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Lanthanum, pieces
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
Lanthanum, rod, 100mm, diameter 6.35mm, cast, 99%
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Lanthanum, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%