Merck
  • Atenolol. A reappraisal of its pharmacological properties and therapeutic use in cardiovascular disorders.
Atenolol. A reappraisal of its pharmacological properties and therapeutic use in cardiovascular disorders.
Drugs (1991-09-01)
A N Wadworth, D Murdoch, R N Brogden
ABSTRACT
Atenolol is a selective beta 1-adrenoceptor antagonist with a duration of activity of at least 24 hours. The scope of therapeutic use of the drug has been expanded and become better defined since it was first reviewed in the Journal in 1979. Atenolol is effective and generally well tolerated in patients with all grades of hypertension. Data from comparative studies show that when administered orally, atenolol reduces blood pressure to a similar extent, and in a similar proportion of patients, as usual therapeutic doses of other beta-adrenoceptor antagonists (such as acebutolol, celiprolol, betaxolol, indenolol, metoprolol, nadolol, pindolol, propranolol, tertatolol), angiotensin converting enzyme (ACE) inhibitors (e.g. captopril, enalapril and lisinopril), calcium antagonists (e.g. amlodipine, diltiazem, felodipine, isradipine, nitrendipine, nifedipine, verapamil), doxazosin, ketanserin and alpha-methyldopa. Atenolol effectively lowers blood pressure in elderly patients with hypertension and in women with hypertension associated with pregnancy, and improves objective and subjective indices in patients with stable angina pectoris. Oral atenolol is used for preventing recurrence of supraventricular arrhythmias once control is achieved by intravenous administration of atenolol. Early intervention with intravenous atenolol followed by oral maintenance therapy reduces infarct recurrence and cardiovascular mortality in patients with known or suspected myocardial infarction. There is also encouraging evidence of reduced mortality from cardiovascular disease during long term therapy with atenolol in patients with hypertension. Atenolol is well tolerated in most patients. Increases in plasma levels of both total triglycerides and very low density lipoprotein (VLDL) triglycerides have accompanied atenolol therapy although the clinical relevance, if any, of longer term metabolic effects has yet to be determined. Its low lipid solubility and limited brain penetration results in a lower incidence of central nervous system effects than that associated with propranolol. After many years of clinical usage atenolol is a well established treatment option in several areas of cardiovascular medicine such as mild to moderate hypertension and stable angina pectoris. Furthermore, it has also shown potential in the treatment of some cardiac arrhythmias and has been associated with reduced cardiovascular mortality in patients with hypertension and in patients with myocardial infarction.
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Atenolol, European Pharmacopoeia (EP) Reference Standard