Merck
Clinical relevance of cimetidine drug interactions.
Drug safety (1992-07-01)
A F Shinn
ABSTRACT
The excellent efficacy and tolerability profiles of H2-antagonists have established these agents as the leading class of antiulcer drugs. Attention has been focused on drug interactions with H2-antagonists as a means of product differentiation and because many patients are receiving multiple drug therapy. The main mechanism of most drug interactions involving cimetidine appears to be inhibition of the hepatic microsomal enzyme cytochrome P450, an effect which may be related to the different structures of H2-antagonists. Ranitidine appears to have less affinity than cimetidine for this system. There have been many published case reports and studies of drug interactions with cimetidine, but many of these have provided pharmacokinetic data only, with little information concerning the clinical significance of these findings. Nevertheless, the coadministration of cimetidine with drugs that have a narrow therapeutic margin (such as theophylline) may potentially result in clinically significant adverse effects. The monitoring of serum concentrations of drugs coadministered with cimetidine may reduce the risk of adverse events but does not abolish the problem. However, for most patients, concomitant administration of cimetidine with drugs possessing a wide therapeutic margin is unlikely to pose a significant problem.
Product Description
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cimetidine
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard