Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%
Iridium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Iridium, foil, 0.025m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%
Iridium, microfoil, disks, 25mm, thinness 0.1μm, specific density 224μg/cm2, permanent mylar 3.5μm support, 99.9%
Iridium, wire reel, 0.2m, diameter 0.15mm, as drawn, 99.9%
Iridium, rod, 25mm, diameter 2.0mm, as drawn, 99.9%
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.5%
Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Iridium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Iridium, wire reel, 0.1m, diameter 0.2mm, as drawn, 99.9%
Iridium, microfoil, disks, 10mm, thinness 0.1μm, specific density 224μg/cm2, permanent mylar 3.5μm support, 99.9%
Iridium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Iridium, wire reel, 0.1m, diameter 0.38mm, as drawn, 99.9%
Iridium, wire reel, 0.05m, diameter 0.8mm, as drawn, 99.9%
Iridium, wire reel, 0.05m, diameter 2.0mm, as drawn, 99.9%
Iridium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Iridium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.9%
Iridium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.9%
Iridium, wire reel, 0.05m, diameter 0.075mm, as drawn, 99.9%
Iridium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Iridium, wire reel, 0.3m, diameter 0.5mm, as drawn, 99.9%
Iridium, wire reel, 0.5m, diameter 0.075mm, as drawn, 99.9%
Iridium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.5%
Iridium, wire reel, 1m, diameter 0.125mm, as drawn, 99.5%
Iridium, wire, straight, 100mm, diameter 0.5mm, as drawn, 99.9%
Iridium, wire reel, 0.5m, diameter 0.15mm, as drawn, 99.9%
Iridium, wire reel, 1m, diameter 0.15mm, as drawn, 99.9%