Merck
  • Isothermal DNA amplification using the T4 replisome: circular nicking endonuclease-dependent amplification and primase-based whole-genome amplification.
Isothermal DNA amplification using the T4 replisome: circular nicking endonuclease-dependent amplification and primase-based whole-genome amplification.
Nucleic acids research (2010-10-06)
Yolanda Schaerli, Viktor Stein, Michelle M Spiering, Stephen J Benkovic, Chris Abell, Florian Hollfelder
ABSTRACT
In vitro reconstitution of the bacteriophage T4 replication machinery provides a novel system for fast and processive isothermal DNA amplification. We have characterized this system in two formats: (i) in circular nicking endonuclease-dependent amplification (cNDA), the T4 replisome is supplemented with a nicking endonuclease (Nb.BbvCI) and a reverse primer to generate a well-defined uniform double-stranded linear product and to achieve up to 1100-fold linear amplification of a plasmid in 1 h. (ii) The T4 replisome with its primase (gp61) can also support priming and exponential amplification of genomic DNA in primase-based whole-genome amplification (T4 pWGA). Low amplification biases between 4.8 and 9.8 among eight loci for 0.3-10 ng template DNA suggest that this method is indeed suitable for uniform whole-genome amplification. Finally, the utility of the T4 replisome for isothermal DNA amplification is demonstrated in various applications, including incorporation of functional tags for DNA labeling and immobilization; template generation for in vitro transcription/translation and sequencing; and colony screening and DNA quantification.
Product Description
GenomiPhi HY Kit, Cytiva 25-6600-22, sufficient for 25 reactions
Genomiphi Hy Kit, Cytiva 25-6600-20, sufficient for 100 reactions
Genomiphi Hy Kit, Cytiva 25-6600-25, sufficient for 1000 reactions