Merck
  • The genetic heterogeneity and mutational burden of engineered melanomas in zebrafish models.
The genetic heterogeneity and mutational burden of engineered melanomas in zebrafish models.
Genome biology (2013-10-24)
Jennifer Yen, Richard M White, David C Wedge, Peter Van Loo, Jeroen de Ridder, Amy Capper, Jennifer Richardson, David Jones, Keiran Raine, Ian R Watson, Chang-Jiun Wu, Jiqiu Cheng, Iñigo Martincorena, Serena Nik-Zainal, Laura Mudie, Yves Moreau, John Marshall, Manasa Ramakrishna, Patrick Tarpey, Adam Shlien, Ian Whitmore, Steve Gamble, Calli Latimer, Erin Langdon, Charles Kaufman, Mike Dovey, Alison Taylor, Andy Menzies, Stuart McLaren, Sarah O'Meara, Adam Butler, Jon Teague, James Lister, Lynda Chin, Peter Campbell, David J Adams, Leonard I Zon, E Elizabeth Patton, Derek L Stemple, P Andy Futreal
ABSTRACT
Melanoma is the most deadly form of skin cancer. Expression of oncogenic BRAF or NRAS, which are frequently mutated in human melanomas, promote the formation of nevi but are not sufficient for tumorigenesis. Even with germline mutated p53, these engineered melanomas present with variable onset and pathology, implicating additional somatic mutations in a multi-hit tumorigenic process. To decipher the genetics of these melanomas, we sequence the protein coding exons of 53 primary melanomas generated from several BRAF(V600E) or NRAS(Q61K) driven transgenic zebrafish lines. We find that engineered zebrafish melanomas show an overall low mutation burden, which has a strong, inverse association with the number of initiating germline drivers. Although tumors reveal distinct mutation spectrums, they show mostly C > T transitions without UV light exposure, and enrichment of mutations in melanogenesis, p53 and MAPK signaling. Importantly, a recurrent amplification occurring with pre-configured drivers BRAF(V600E) and p53-/- suggests a novel path of BRAF cooperativity through the protein kinase A pathway. This is the first analysis of a melanoma mutational landscape in the absence of UV light, where tumors manifest with remarkably low mutation burden and high heterogeneity. Genotype specific amplification of protein kinase A in cooperation with BRAF and p53 mutation suggests the involvement of melanogenesis in these tumors. This work is important for defining the spectrum of events in BRAF or NRAS driven melanoma in the absence of UV light, and for informed exploitation of models such as transgenic zebrafish to better understand mechanisms leading to human melanoma formation.
Product Description
Genomiphi Hy Kit, Cytiva 25-6600-20, sufficient for 100 reactions
Genomiphi Hy Kit, Cytiva 25-6600-25, sufficient for 1000 reactions
GenomiPhi HY Kit, Cytiva 25-6600-22, sufficient for 25 reactions