Merck
  • Bioequivalence study of two commercial amoxicillin suspension formulations in healthy human volunteers.
Bioequivalence study of two commercial amoxicillin suspension formulations in healthy human volunteers.
International journal of clinical pharmacology and therapeutics (2014-04-24)
Gilson C N Franco, Sinvaldo Baglie, Ana P B Ruenis, Luiz M Franco, Karina Cogo, Yoko Oshima-Franco, Paulo Silva, Francisco C Groppo, Pedro Luiz Rosalen
ABSTRACT
To compare the pharmacokinetic profiles and to evaluate the bioequivalence of two commercial amoxicillin suspension formulations (500 mg/5 mL AMOXIL®, reference formulation and AMOXI-PED®, test formulation) in healthy Brazilian volunteers. Under fasting condition, 25 volunteers (13 males and 12 females) were included in this randomized, open-label, two-period crossover (1-week washout interval) bioequivalence study. Blood samples were collected at pre-dose (0 hour) and 0.5, 1, 1.33, 1.66, 2, 2.5, 3, 4, 6, 8, and 12 hours after drug ingestion. Pharmacokinetic parameters (Cmax, tmax, t1/2, AUC0-tlast, and AUC0-∞) were calculated from plasma concentrations for both formulations in each subject. Arithmetic mean values of the pharmacokinetic parameters were: Cmax = 12.004 (± 2.824) μg×mL-1; tmax = 1.118 (± 0.396) h; t1/2 = 1.226 (± 0.179) h; AUC0-tlast = 29.297 (± 6.007) μg×h×mL-1; and AUC0-∞ = 29.299 (± 6.007) μg×h×mL-1 for reference formulation and Cmax = 11.456 (± 2.825) μg×mL-1; tmax = 1.331 (± 0.509) h; t1/2 = 1.141 (± 0.133) h; AUC0-tlast = 28.672 (± 5.778) μg×h×mL-1; and AUC0-∞ = 28.693 (± 5.796) μg×h×mL-1 for test formulation. The confidence intervals (90% CI) for reference and test formulations were, respectively, 90.74 - 100.46% for Cmax and 93.62 - 103.61% for AUC0-t. Based on the results, both formulations of amoxicillin evaluated in this study were considered bioequivalent according to FDA and ANVISA/Brazil criteria.
Product Description
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
Amoxicillin trihydrate for performance verification, European Pharmacopoeia (EP) Reference Standard
Supelco
Amoxicillin trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Amoxicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Amoxicillin trihydrate, European Pharmacopoeia (EP) Reference Standard