Merck
  • Band broadening along gradient reversed phase columns: a potential gain in resolution factor.
Band broadening along gradient reversed phase columns: a potential gain in resolution factor.
Journal of chromatography. A (2014-04-17)
Fabrice Gritti, Georges Guiochon
ABSTRACT
The advantages of using columns packed with stationary phases having a composition gradient so that retention factors increase toward the outlet and eluting them with an isocratic mobile phase may provide increased peak resolution in liquid chromatography. This approach is discussed from a theoretical viewpoint. The retention factor is assumed to increase linearly along the column. The peak width can be predicted under three different conditions: (1) the possible band compression is neglected (the Giddings model); (2) band compression is taken into account (the Poppe model); and (3) both band compression and extra-column effects are considered in the calculations. The impact of a stationary phase gradient on the resolution of a pair of compound difficult to separate (constant selectivity α=1.05) is illustrated for a 3mm×100mm silica monolithic column of the second generation (porosity ϵt=0.85, plate height H=6μm) operated with a new generation of liquid chromatograph (pre- and post-column volume variances of 10 and 5μL(2), respectively). The results show that the resolution factor RS becomes optimum for a specific positive value of the retention gradient along the column. This optimum depends on the retention factor at the column inlet.
Sigma-Aldrich
Silica
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O