Merck
  • Candida albicans SOD5 represents the prototype of an unprecedented class of Cu-only superoxide dismutases required for pathogen defense.
Candida albicans SOD5 represents the prototype of an unprecedented class of Cu-only superoxide dismutases required for pathogen defense.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-09)
Julie E Gleason, Ahmad Galaleldeen, Ryan L Peterson, Alexander B Taylor, Stephen P Holloway, Jessica Waninger-Saroni, Brendan P Cormack, Diane E Cabelli, P John Hart, Valeria Cizewski Culotta
ABSTRACT
The human fungal pathogens Candida albicans and Histoplasma capsulatum have been reported to protect against the oxidative burst of host innate immune cells using a family of extracellular proteins with similarity to Cu/Zn superoxide dismutase 1 (SOD1). We report here that these molecules are widespread throughout fungi and deviate from canonical SOD1 at the primary, tertiary, and quaternary levels. The structure of C. albicans SOD5 reveals that although the β-barrel of Cu/Zn SODs is largely preserved, SOD5 is a monomeric copper protein that lacks a zinc-binding site and is missing the electrostatic loop element proposed to promote catalysis through superoxide guidance. Without an electrostatic loop, the copper site of SOD5 is not recessed and is readily accessible to bulk solvent. Despite these structural deviations, SOD5 has the capacity to disproportionate superoxide with kinetics that approach diffusion limits, similar to those of canonical SOD1. In cultures of C. albicans, SOD5 is secreted in a disulfide-oxidized form and apo-pools of secreted SOD5 can readily capture extracellular copper for rapid induction of enzyme activity. We suggest that the unusual attributes of SOD5-like fungal proteins, including the absence of zinc and an open active site that readily captures extracellular copper, make these SODs well suited to meet challenges in zinc and copper availability at the host-pathogen interface.
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper, tube, 1000mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper, sphere, 1000pcs, diameter 3.0mm, precision sphere grade 200
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Copper, tube, 100mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, sphere, 100pcs, diameter 3.0mm, precision sphere grade 200
Copper, rod, 50mm, diameter 12.7mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 2m coil, thickness 0.7mm, half hard, 99.95+%
Copper, wire reel, 50m, diameter 0.25mm, as drawn, 99.99+%
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Copper, wire reel, 500m, diameter 0.125mm, annealed, 99.9%
Copper, wire reel, 2m, diameter 0.75mm, as drawn, 99.98+%
Copper, wire reel, 20m, diameter 0.5mm, as drawn, 99.999%
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Copper, wire reel, 500m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 250m, diameter 0.04mm, annealed, 99.9%