- Thermodynamic and structural characterization of the specific binding of Zn(II) to human protein DJ-1.
Thermodynamic and structural characterization of the specific binding of Zn(II) to human protein DJ-1.
Biochemistry (2014-04-05)
Shinya Tashiro, Jose M M Caaveiro, Chun-Xiang Wu, Quyen Q Hoang, Kouhei Tsumoto
PMID24697266
ABSTRACT
Mutations of DJ-1 cause familial Parkinson's disease (PD), although the role of DJ-1 in PD remains unresolved. Very recent reports have shown that DJ-1 interacts with copper ions. This evidence opens new avenues to understanding the function of DJ-1 and its role in PD. Herein, we report that Zn(II) binds to DJ-1 with great selectivity among the other metals examined: Mn(II), Fe(II), Co(II), Ni(II), and Cu(II). High-resolution X-ray crystallography (1.18 Å resolution) shows Zn(II) is coordinated to the protein by the key residues Cys106 and Glu18. These results suggest that DJ-1 may be regulated and/or stabilized by Zn(II).
Product Description
Zinc, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 150x150mm, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 100x100mm, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 100x100mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 2m coil, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, not light tested, 25x25mm, thickness 0.015mm, 99.9%
Zinc, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.02mm, 99.9%
Zinc, foil, not light tested, 100x100mm, thickness 0.01mm, 99.9%
Zinc, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%