Merck
  • Analysis of gene expression changes, caused by exposure to nitrite, in metabolic and antioxidant enzymes in the red claw crayfish, Cherax quadricarinatus.
Analysis of gene expression changes, caused by exposure to nitrite, in metabolic and antioxidant enzymes in the red claw crayfish, Cherax quadricarinatus.
Ecotoxicology and environmental safety (2014-04-01)
Qichen Jiang, Wenyi Zhang, Hongyue Tan, Dongmei Pan, Yuanhao Yang, Qian Ren, Jiaxin Yang
ABSTRACT
We evaluated the effect of acute exposure to nitrite on expression of antioxidant and metabolic enzyme genes in gill tissue of advanced juvenile Cherax quadricarinatus. A 48h nitrite exposure was conducted, using four test concentrations (NO2-N=0.5, 1, 1.5 and 2mg L(-1)) plus a control group. The relative mRNA expression of mitochondrial manganese superoxide dismutase (mMnSOD), cytosolic MnSOD (cMnSOD), extracellular copper/zinc SOD (exCu/ZnSOD), catalase (CAT), glutathione S-transferase (GST), arginine kinase (AK), glutamate dehydrogenase (GDH), mitochondrial malate dehydrogenase (mMDH), Na(+)/K(+)-ATPase α-subunit and phosphoenolpyruvate carboxykinase (PEPCK) in gill tissue was measured. Significantly increased mRNA expression was observed for all the antioxidant enzymes after 12 and 24h. After 48h, they all decreased at high nitrite concentrations. The gene expression levels of AK, GDH, mMDH and Na(+)/K(+)-ATPase α-subunit showed similar trends as the antioxidant enzymes. Significant depression of gene expression levels of PEPCK occurred throughout the experimental time at high nitrite concentrations. The results indicated that nitrite could induce oxidative and metabolic stress in C. quadricarinatus, in a time dependent manner, which suggests they could be helpful in predicting sublethal nitrite toxicity and useful in environmental monitoring studies.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard