Merck
Copper-mediated trifluoromethylthiolation of α-diazoesters.
Organic letters (2014-03-26)
Mingyou Hu, Jian Rong, Wenjun Miao, Chuanfa Ni, Yongxin Han, Jinbo Hu
ABSTRACT
A novel Cu-mediated trifluoromethylthiolation of diazo compounds has been developed that provides a convenient synthetic route for the efficient α-trifluoromethylthiolation of simple esters under mild reaction conditions. The reaction is typically carried out at room temperature, and water could be used to promote the reaction.
Product Description
USP
Residual Solvent Class 2 - N,N-Dimethylacetamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylacetamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylacetamide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for HPLC, ≥99.9%