Merck
  • Efficacy of sonazoid (perflubutane) for contrast-enhanced ultrasound in the differentiation of focal breast lesions: phase 3 multicenter clinical trial.
Efficacy of sonazoid (perflubutane) for contrast-enhanced ultrasound in the differentiation of focal breast lesions: phase 3 multicenter clinical trial.
AJR. American journal of roentgenology (2014-03-26)
Yukio Miyamoto, Toshikazu Ito, Etsuo Takada, Kiyoka Omoto, Toshiko Hirai, Fuminori Moriyasu
ABSTRACT
The objective of our study was to compare the efficacy of contrast-enhanced ultrasound (CEUS) using the ultrasound contrast agent Sonazoid (perflubutane) with unenhanced ultrasound and supplementary contrast-enhanced MRI in the differential diagnosis (benign vs malignant) of focal breast lesions. The safety of Sonazoid was also assessed in this study. A total of 127 patients with focal breast lesions were enrolled in this study at five centers in Japan. Three reviewers who were blinded to the patient characteristics independently assessed the ultrasound images and MR images in a randomized sequence. The accuracy, sensitivity, and specificity of CEUS, unenhanced ultrasound, and supplementary contrast-enhanced MRI for the differential diagnosis were compared using generalized estimating equation analyses. Diagnostic confidence was also assessed. The accuracy of CEUS was significantly higher than that of unenhanced ultrasound (87.2% vs 65.5%, respectively; p < 0.001). In addition, CEUS showed significantly higher specificity, although the improvement in sensitivity was not statistically significant. The accuracy and specificity were significantly higher with CEUS than with contrast-enhanced MRI, but the improvement in sensitivity was not statistically significant. The area under the curve in a receiver operating characteristic analysis was significantly greater with CEUS than with unenhanced ultrasound. The incidence of adverse events was 11.4% and the incidence of adverse drug reactions was 3.3%. All adverse drug reactions were mild. CEUS using Sonazoid was confirmed to be superior to unenhanced ultrasound for the differential diagnosis (benign vs malignant) of focal breast lesions in terms of diagnostic accuracy with no serious adverse reactions.
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, rod, 1000mm, diameter 30mm, as drawn, 98+%
Iron, rod, 150mm, diameter 5.0mm, as drawn, 99.99+%
Iron, foil, 100x100mm, thickness 0.15mm, hard, 99.5%
Iron, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, rod, 100mm, diameter 4.8mm, as drawn, 98+%
Iron, foil, not light tested, 50x50mm, thickness 0.002mm, 99.85%
Iron, rod, 100mm, diameter 100mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 500mm, diameter 19mm, as drawn, soft ingot 99.8%
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Iron, rod, 1000mm, diameter 12.7mm, as drawn, 98+%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, rod, 500mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 150x150mm, thickness 0.009mm, 99.85%
Iron, rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Iron, rod, 1000mm, diameter 19.0mm, as drawn, 98+%
Iron, foil, 150x150mm, thickness 0.38mm, hard, 99.5%
Iron, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.01mm, polyester insulation
Iron, rod, 500mm, diameter 50mm, as drawn, 98+%
Iron, rod, 200mm, diameter 12.7mm, as drawn, 99.95%
Iron, rod, 200mm, diameter 9.5mm, as drawn, 98+%
Iron, foil, 25x25mm, thickness 1.83mm, Quarter hard, 99.5%