Merck
  • Bevacizumab combined with chemotherapy for platinum-resistant recurrent ovarian cancer: The AURELIA open-label randomized phase III trial.
Bevacizumab combined with chemotherapy for platinum-resistant recurrent ovarian cancer: The AURELIA open-label randomized phase III trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-03-19)
Eric Pujade-Lauraine, Felix Hilpert, Béatrice Weber, Alexander Reuss, Andres Poveda, Gunnar Kristensen, Roberto Sorio, Ignace Vergote, Petronella Witteveen, Aristotelis Bamias, Deolinda Pereira, Pauline Wimberger, Ana Oaknin, Mansoor Raza Mirza, Philippe Follana, David Bollag, Isabelle Ray-Coquard
ABSTRACT
In platinum-resistant ovarian cancer (OC), single-agent chemotherapy is standard. Bevacizumab is active alone and in combination. AURELIA is the first randomized phase III trial to our knowledge combining bevacizumab with chemotherapy in platinum-resistant OC. Eligible patients had measurable/assessable OC that had progressed < 6 months after completing platinum-based therapy. Patients with refractory disease, history of bowel obstruction, or > two prior anticancer regimens were ineligible. After investigators selected chemotherapy (pegylated liposomal doxorubicin, weekly paclitaxel, or topotecan), patients were randomly assigned to single-agent chemotherapy alone or with bevacizumab (10 mg/kg every 2 weeks or 15 mg/kg every 3 weeks) until progression, unacceptable toxicity, or consent withdrawal. Crossover to single-agent bevacizumab was permitted after progression with chemotherapy alone. The primary end point was progression-free survival (PFS) by RECIST. Secondary end points included objective response rate (ORR), overall survival (OS), safety, and patient-reported outcomes. The PFS hazard ratio (HR) after PFS events in 301 of 361 patients was 0.48 (95% CI, 0.38 to 0.60; unstratified log-rank P < .001). Median PFS was 3.4 months with chemotherapy alone versus 6.7 months with bevacizumab-containing therapy. RECIST ORR was 11.8% versus 27.3%, respectively (P = .001). The OS HR was 0.85 (95% CI, 0.66 to 1.08; P < .174; median OS, 13.3 v 16.6 months, respectively). Grade ≥ 2 hypertension and proteinuria were more common with bevacizumab. GI perforation occurred in 2.2% of bevacizumab-treated patients. Adding bevacizumab to chemotherapy statistically significantly improved PFS and ORR; the OS trend was not significant. No new safety signals were observed.
Product Description
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard