Merck
  • Manganese in exhaled breath condensate: a new marker of exposure to welding fumes.
Manganese in exhaled breath condensate: a new marker of exposure to welding fumes.
Toxicology letters (2014-02-11)
Sébastien Hulo, Nathalie Chérot-Kornobis, Mike Howsam, Sébastien Crucq, Virginie de Broucker, Annie Sobaszek, Jean-Louis Edme
ABSTRACT
To evaluate manganese in exhaled breath condensate (Mn-EBC) as an indicator of exposure to fumes from metal inert gas welding process. We collected EBC and urine from 17 welders and 16 unexposed control subjects after 5 days exposure. Concentrations of manganese (Mn), nickel (Ni), iron (Fe) and chromium (Cr) were measured in EBC and urine samples and correlated with cumulative exposure indices for the working week (CIW) and for the total welding years (WY), based on duration of welding activity and atmospheric metal measurements. Concentrations of Mn and Ni in EBC were significantly higher among welders than controls whereas this difference was not significant for Mn in urine. Levels of Mn and Ni in EBC were not correlated with their respective levels in urine. The linear regressions found significant positive coefficients between Mn-EBC, Ni-EBC, Ni-U and Cr-U concentrations and the cumulative exposure indices. Taking into account tobacco use, statistical analysis showed the same trends except for the relationship between Mn-U and CIW. This pilot study showed that Mn-EBC, as well as Ni-EBC, can serve as reliable indices of occupational exposure to welding fumes and provide complimentary toxicokinetic information to that provided by urine analyses.
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Nickel, foil, not light tested, 100x100mm, thickness 0.002mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.0125mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.025mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.20mm, annealed, 99%
Nickel, foil, 10m coil, thickness 0.05mm, coil width 2mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.075mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.125mm, annealed, 99.98%
Nickel, wire reel, 100m, diameter 0.5mm, as drawn, 99%
Nickel, microfoil, disks, 25mm, thinness 1.0μm, specific density 895.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, foil, 50x50mm, thickness 1.0mm, annealed, 99.98%
Nickel, foil, 2m coil, thickness 0.3mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 3.6mm, inside diameter 3.5mm, wall thickness 0.05mm, hard, 99.5%
Nickel, foil, 150x300mm, thickness 0.5mm, annealed, 99.98%
Nickel, wire reel, 10m, diameter 0.125mm, hard, 99.98%
Nickel, foil, not light tested, 50x50mm, thickness 0.007mm, 99.95%
Nickel, tube, 500mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, hard, 99.5%
Nickel, insulated wire, 20m, conductor diameter 0.025mm, insulation thickness 0.0035mm, polyimide insulation
Nickel, tube, 500mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%
Nickel, foil, 25x25mm, thickness 3.2mm, as rolled, 99.99+%