Merck
  • Endothelin-1 regulates H⁺-ATPase-dependent transepithelial H⁺ secretion in zebrafish.
Endothelin-1 regulates H⁺-ATPase-dependent transepithelial H⁺ secretion in zebrafish.
Endocrinology (2014-01-16)
Ying-Jey Guh, Yung-Che Tseng, Chao-Yew Yang, Pung-Pung Hwang
ABSTRACT
Endothelin-1 (EDN1) is an important regulator of H⁺ secretion in the mammalian kidney. EDN1 enhances renal tubule H⁺-ATPase activity, but the underlying mechanism remains unclear. To further elucidate the role of EDN1 in vertebrates' acid-base regulation, the present study used zebrafish as the model to examine the effects of EDN1 and its receptors on transepithelial H⁺ secretion. Expression of EDN1 and one of its receptors, EDNRAa, was stimulated in zebrafish acclimated to acidic water. A noninvasive scanning ion-selective electrode technique was used to show that edn1 overexpression enhances H⁺ secretion in embryonic skin at 3 days post fertilization. EDNRAa loss of function significantly decreased EDN1- and acid-induced H⁺ secretion. Abrogation of EDN1-enhanced H⁺ secretion by a vacuolar H⁺-ATPase inhibitor (bafilomycin A1) suggests that EDN1 exerts its action by regulating the H⁺-ATPase-mediated H⁺ secretion. EDN1 does not appear to affect H⁺ secretion through either altering the abundance of H⁺-ATPase or affecting the cell differentiation of H⁺-ATPase-rich ionocytes, because the reduction in secretion upon ednraa knockdown was not accompanied by decreased expression of H⁺-ATPase or reduced H⁺-ATPase-rich cell density. These findings provide evidence that EDN1 signaling is involved in acid-base regulation in zebrafish and enhance our understanding of EDN1 regulation of transepithelial H⁺ secretion in vertebrates.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Hydrogen ionophore I, Selectophore, function tested
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Tridodecylamine, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water