Merck
  • Nitrogen-bisphosphonate therapy is linked to compromised coenzyme Q10 and vitamin E status in postmenopausal women.
Nitrogen-bisphosphonate therapy is linked to compromised coenzyme Q10 and vitamin E status in postmenopausal women.
The Journal of clinical endocrinology and metabolism (2014-01-16)
Shirin Kalyan, Patricia Huebbe, Tuba Esatbeyoglu, Petra Niklowitz, Hélène C F Côté, Gerald Rimbach, Dieter Kabelitz
ABSTRACT
Nitrogen-bisphosphonates (N-BPs) are the most widely used drugs for bone fragility disorders. Long-term or high-dose N-BP use is associated with unusual serious side effects such as osteonecrosis of the jaw, musculoskeletal pain, and atypical fractures of long bones. It has escaped notice that the pathway N-BPs block is central for the endogenous synthesis of coenzyme Q10, an integral enzyme of the mitochondrial respiratory chain and an important lipid-soluble antioxidant. Our objective was to assess the coenzyme Q10 and antioxidant status in relation to N-BP exposure in women with postmenopausal osteoporosis. Seventy-one postmenopausal women (age, 73.5 ± 5.5 y) with osteoporosis and no other malignancy were included in this cross-sectional study. Seventeen were treatment naive, 27 were on oral N-BP, and 27 were on i.v. N-BP. Vitamin E γ-tocopherol levels (μmol/mL) were significantly reduced in N-BP users [oral, H(2) = 18.5, P = .02; i.v., H(2) = 25.2, P < .001; mean rank comparisons after Kruskal-Wallis test). Length of time (days) of N-BP exposure, but not age, was inversely associated with the coenzyme Q10/cholesterol ratio (μmol/mol) (β = -0.27; P = .025), which was particularly low for those on i.v. N-BP (mean difference = -35.0 ± 16.9; 95% confidence interval, -65.2 to -4.9; P = .02). The degree of N-BP exposure appears related to compromised coenzyme Q10 status and vitamin E γ-tocopherol levels in postmenopausal women with osteoporosis. This phenomenon may link to certain adverse N-BP-associated effects. Confirmation of this would suggest that therapeutic supplementation could prevent or reverse certain complications of long-term N-BP therapy for at-risk individuals.
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
USP
Ubidecarenone, United States Pharmacopeia (USP) Reference Standard
USP
Ubidecarenone for System Suitability, United States Pharmacopeia (USP) Reference Standard
Supelco
Coenzyme Q10, analytical standard
Ubidecarenone, European Pharmacopoeia (EP) Reference Standard
Ubidecarenone for system suitability, European Pharmacopoeia (EP) Reference Standard