Merck
  • Cadmium phytoavailability to rice (Oryza sativa L.) grown in representative Chinese soils. A model to improve soil environmental quality guidelines for food safety.
Cadmium phytoavailability to rice (Oryza sativa L.) grown in representative Chinese soils. A model to improve soil environmental quality guidelines for food safety.
Ecotoxicology and environmental safety (2014-01-15)
Muhammad T Rafiq, Rukhsanda Aziz, Xiaoe Yang, Wendan Xiao, Muhammad K Rafiq, Basharat Ali, Tingqiang Li
ABSTRACT
Food chain contamination by cadmium (Cd) is globally a serious health concern resulting in chronic abnormalities. Rice is a major staple food of the majority world population, therefore, it is imperative to understand the relationship between the bioavailability of Cd in soils and its accumulation in rice grain. Objectives of this study were to establish environment quality standards for seven different textured soils based on human dietary toxicity, total Cd content in soils and bioavailable portion of Cd in soil. Cadmium concentrations in polished rice grain were best related to total Cd content in Mollisols and Udic Ferrisols with threshold levels of 0.77 and 0.32mgkg(-1), respectively. Contrastingly, Mehlich-3-extractable Cd thresholds were more suitable for Calcaric Regosols, Stagnic Anthrosols, Ustic Cambosols, Typic Haplustalfs and Periudic Argosols with thresholds values of 0.36, 0.22, 0.17, 0.08 and 0.03mgkg(-1), respectively. Stepwise multiple regression analysis indicated that phytoavailability of Cd to rice grain was strongly correlated with Mehlich-3-extractable Cd and soil pH. The empirical model developed in this study explains the combined effects of soil properties and extractable soil Cd content on the phytoavailability of Cd to polished rice grain. This study indicates that accumulation of Cd in rice is influenced greatly by soil type, which should be considered in assessment of soil safety for Cd contamination in rice. This investigation concluded that the selection of proper soil type for food crop production can help us to avoid the toxicity of Cd in our daily diet.
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, wire reel, 0.1m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.7+%
Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 0.5m coil, thickness 0.5mm, as rolled, 100%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.95+%
Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, wire reel, 0.5m, diameter 1.0mm, hard, 99.999%
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.99+%