Merck
Lead and cadmium contaminations in feathers of heron and egret chicks.
Environmental monitoring and assessment (2014-01-11)
Jungsoo Kim, Jong-Min Oh
ABSTRACT
In birds, concentrations of heavy metal contaminants were associated with their diet and environmental quality. This study presents concentrations of lead and cadmium in feathers and the relationship between concentrations in the feather and stomach content of three species of heron and egret chicks. Cadmium concentrations were higher in the Little Egret (geomean 0.75 μg/g dry weight) and Black-crowned Night Heron (geomean 0.88 μg/g dry weight) chicks than in Grey Heron chicks (geomean 0.48 μg/g dry weight). Lead and cadmium concentrations had significant correlations between feathers and stomach contents of heron and egret chicks. The mean lead and cadmium concentrations in feathers were within the background levels for wild birds. Heron and egret species forage around breeding colony (4-7 km). Lead and cadmium concentrations in heron and egret chicks were moderately associated with those of prey concentrations. These results suggest that contaminant concentrations such as lead and cadmium in feathers of heron and egret chicks can reflect local heavy metal contaminations and serve as a great biological indicator for monitoring heavy metal contamination. In addition, the feathers can be a routine monitoring tool over time and space without harming the wild birds.
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.025mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, light tested, 50x50mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.015mm, 99.7+%